Skip to Content
Merck
All Photos(1)

Documents

MAB1946-IM

Sigma-Aldrich

Anti-Nidogen 1 Antibody, clone ELM1

clone ELM1, from rat

Synonym(s):

Nidogen-1, NID-1, Entactin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ELM1, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Nid1(18073)

General description

Nidogen-1 (UniProt: P10493; also known as NID-1, Entactin) is encoded by the Nid1 (also known as Ent) gene (Gene ID: 18073) ins murine species. Nidogen-1 is a highly conserved sulfated monomeric glycoprotein that is widely distributed in basement membranes and tightly associates with laminin. It is post-translationally N- and O-glycosylated. It is synthesized with a 28 aa signal peptide that is subsequently cleaved to produce the mature form. Nidogen -1 contains six EGF-like domains, four LDL-receptor class B repeats, one NIDO domain, one nidogen G2 barrel domain, and one thyroglobulin type 1 domain. Nidogen-1 shares about 50% homology with Nidigen-2 and shares similar adhesive properties. There is also a high degree of homology between murine and human Nidogen-1. Nidogen is expressed during embryonic and fetal development exclusively in fully developed basement membranes of the ectoderm and is not expressed in the developing endodermal basement membrane or in membranes disrupted during mesoderm formation.

Specificity

Clone ELM1 is a rat monoclonal antibody that detects Nidogen-1 in murine species.

Immunogen

Purified protein from basement membranes of mouse Engelbreth-Holm-Swarm (EHS) tumor.

Application

Anti-Nidogen 1, clone ELM1, Cat. No. MAB1946-I, is a rat monoclonal antibodythat detects Nidogen-1 and has been tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Western Blotting Analysis: A representative lot detected Nidogen-1 in Western Blotting applications (Arends, F., et. al. (2015). PLoS One. 10(2):e0118090).

Immunocytochemistry Analysis: A representative lot detected Nidogen-1 in Immunocytochemistry applications (Kick, K., et. al. (2016). Arterioscler Thromb Vasc Biol. 36(12):2346-2357).

Immunohistochemistry Analysis: A representative lot detected Nidogen-1 in Immunohistochemistry applications (Chang, Y.T., et. al. (2016). J Clin Invest. 126(2):721-31).

Quality

Evaluated by Immunohistochemistry in mouse kidney.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Nidogen-1 in mouse kidney tissue.

Target description

136.54 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service