Skip to Content
Merck
All Photos(1)

Key Documents

HX0293

Supelco

Hexane

≥99.0% (n-hexane basis)

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

n-Hexane 99%, 99%

vapor density

~3 (vs air)

Quality Level

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥99.0% (n-hexane basis)

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

impurities

≤0.05% Water (H2O)

color

APHA: ≤10

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.659 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

99%

Analysis Note

Assay (n-Hexane): 99.0% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Water (H2O): 0.05% max

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service