Skip to Content
Merck
All Photos(3)

Key Documents

HTS017M

Sigma-Aldrich

ChemiSCREEN C5aR Membrane Preparation, recombinant

Human C5aR GPCR membrane preparation for Radioligand binding Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

C5a is a proinflammatory peptide generated through activation of the complement system, and is more potent than the other anaphylatoxins, C4a and C3a, in activating peripheral blood leukocytes (Gerard & Gerard, 1994). It can actively participate in the regulation of local cytokine network by stimulating production of proinflammatory cytokines (Buchner et al., 1995; Hsu, et al., 1999). The proinflammatory effects of C5a are mediated through binding to a specific 7-TM chemoattractant receptor, C5aR (CD88). Binding of C5a to phagocyte C5aR induces chemotaxis, production of superoxide anions, and release of degradative enzymes. Pharmacologic or genetic disruption of C5a/C5aR interaction reduces sepsis (Riedemann et al., 2003), immune complex-induced lung disease (Shushakova et al., 2002), and Arthrogen-induced arthritis (Grant et al., 2002) in experimental models. Chemicon′s cloned human C5a receptor membrane preparations are ideal tools for screening for antagonists of C5a/C5aR interactions. The C5aR Membrane Preps bind specifically to 125I-labeled ligand C5a, with Kd of 0.59 nM. With 5 μg membrane per well and 0.1 nM 125I-labeled C5a, more than 15-fold signal:background was obtained.
Full-length human C5aR cDNA

Application

Radioligand binding assay (See images & graphs link)

Biochem/physiol Actions

GPCR Class: A
Protein Target: C5aR
Target Sub-Family: Anaphylotoxin

Quality

SPECIFICATIONS: Bmax for [125I]C5a binding: 9.7 pmol/mg protein
Kd for [125I] C5a binding: ~ 0.59 nM

Physical form

Membranes were adjusted to 0.5 mg protein/ml in 50 mM Tris pH 7.4, 10% glycerol and 1% BSA, rapidly frozen, and stored at -80°C. One vial contains 1000 units, where a unit is the amount of membrane that will yield greater than 1000 cpm specific binding of I-125 labeled C5a (0.1 nM input concentration).

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service