Skip to Content
Merck
All Photos(1)

Key Documents

CP70

Sigma-Aldrich

Anti-Fibronectin (Ab-3) Mouse mAb (FBN11)

liquid, clone FBN11, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

FBN11, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)
mouse ... Fn1(14268)
rat ... Fn1(25661)

General description

Anti-Fibronectin (Ab-3), mouse monoclonal, clone FBN11, recognizes dimeric fibronectin in SWI56 cells and normal kidney. It is validated for use in WB, IF, IP & IHC (frozen and paraffin sections).
Protein G purified mouse monoclonal antibody. Recognizes the ~440 kDa disulfide-linked, dimeric fibronectin.
Recognizes the ~440 kDa disulfide-linked, dimeric fibronectin in SWI56 cells and normal kidney tissue.

Immunogen

recombinant human fibronectin

Application

Frozen Sections (0.5-1 µg/ml)

Immunoblotting (0.5-1 µg/ml)

Immunofluorescence (see comments)

Immunoprecipitation (2 µg/mg protein lysate)

Paraffin Sections (0.5-1 µg/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Positive Control
SW 156 cells or normal kidney tissue

Other Notes

Clark, R.A. 2001. Ann. N.Y. Acad. Sci.936, 355.
Armstrong, P.B. and Armstrong, M.T. 2000. Biochim. Biophys. Acta1470, O9.
Barilla, M.L. and Carsons, S.E. 2000. Semin. Arthritis Rheum.29, 252.
Yamada, K.M. 1989. In Fibronectin, Mosher, D.F., ed., p. 47.
Rouslahti, E. and Pierschbacher, M.D. 1986. Cell44, 517.
Pytela, R., et al. 1985. Cell40, 191.
Exhibits staining of the connective tissue matrix when used in immunohistochemistry. This antibody is suitable for both reduced and non-reduced protein by immunoblotting. For staining of formalin-fixed tissues, boil in 10 mM citrate buffer, pH 6.0, for 10-20 min, followed by cooling at room temperature for 20 min. This antibody has also been reported to work for immunofluorescence. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ismaïl Hendaoui et al.
Matrix biology : journal of the International Society for Matrix Biology, 40, 46-53 (2014-09-10)
Whisker follicles have multiple stem cell niches, including epidermal stem cells in the bulge as well as neural crest-derived stem cells and mast cell progenitors in the trabecular region. The neural crest-derived stem cells are a pool of melanocyte precursors.
R Schaal-Jensen et al.
Journal of thrombosis and haemostasis : JTH, 5(10), 2070-2078 (2007-07-26)
In cynomolgus monkeys, suprapharmacological doses of clotting recombinant factor XIII (rFXIII) cause a generalized coagulopathy, associated with formation of circulating high molecular weight protein complexes (HMEX). HMEX consist of plasma protein substrates cross-linked by FXIII transglutaminase activity. To characterize HMEX
Natasha Arora et al.
Development (Cambridge, England), 144(6), 1128-1136 (2017-02-09)
Temporal manipulation of the in vitro environment and growth factors can direct differentiation of human pluripotent stem cells into organoids - aggregates with multiple tissue-specific cell types and three-dimensional structure mimicking native organs. A mechanistic understanding of early organoid formation
Peipei Li et al.
Cell death discovery, 7(1), 59-59 (2021-03-25)
Sirtuin 1 (SIRT1) is a nicotinamide adenine dinucleotide (NAD+)-dependent deacetylase belonging to class III histone deacetylases. Previous studies have shown that SIRT1 is involved in kidney physiology regulation and protects the kidney from various pathological factors. However, the underlying mechanisms
Yu-Jen Chiu et al.
International journal of oncology, 52(5), 1465-1478 (2018-03-24)
Osteosarcoma is the most common primary malignancy of the bone and is characterized by local invasion and distant metastasis. Over the past 20 years, long-term outcomes have reached a plateau even with aggressive therapy. Overexpression of insulin-like growth factor 1 receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service