ACR5000PG
Protein G Reagent Kit
for antibody purification
Synonym(s):
Protein purification kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24
Recommended Products
product line
Amicon®
shipped in
wet ice
Related Categories
General description
Protein G is a bacterial cell wall protein produced by G Streptococci that binds to immunoglobulin (IgG). It is a single polypeptide chain with minimal carbohydrate modifications. Native protein G consists of two IgG binding domains and binding sites for cell surface and albumin. The Amicon® Pro Affinity Concentration Kit - Protein G is developed for maximal binding and elution of Ig species.
Application
Amicon® Pro Affinity Concentration Kit - Protein G is ideal for antibody purification.
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Met. Corr. 1
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service