Skip to Content
Merck
All Photos(1)

Key Documents

8.43075

Sigma-Aldrich

4-Nitrophenyl acetate

for synthesis

Synonym(s):

4-Nitrophenyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO4
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352108
EC Index Number:
212-593-5
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

mp

77-79 °C

transition temp

flash point >110 °C

density

1.244 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)OC(=O)C

InChI

1S/C8H7NO4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3

InChI key

QAUUDNIGJSLPSX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 77 °C
Melting range (upper value): ≤ 79 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service