Skip to Content
Merck
All Photos(1)

Key Documents

8.20430

Sigma-Aldrich

2,6-Dichloroaniline

for synthesis

Synonym(s):

2,6-Dichloroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,6-(Cl)2C6H3NH2
CAS Number:
Molecular Weight:
162.02
MDL number:
UNSPSC Code:
12352116
EC Index Number:
210-160-5

vapor pressure

<0.5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

>600 °C

bp

97 °C/7 hPa

mp

35-38 °C

transition temp

flash point 118 °C

density

1.275 g/cm3 at 30 °C

InChI

1S/C6H5Cl2N/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2

InChI key

JDMFXJULNGEPOI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 35 °C
Melting range (upper value): ≤ 38 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service