Skip to Content
Merck
All Photos(1)

Key Documents

8.20061

Sigma-Aldrich

2-Aminoethyl dihydrogen phosphate

for synthesis

Synonym(s):

2-Aminoethyl dihydrogen phosphate, Colamine phosphoric acid, Ethanolamine dihydrogen phosphate, 2-Hydroxyethylammonium dihydrogen phosphate, O-Phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2OPO(OH)2
CAS Number:
Molecular Weight:
141.06
MDL number:
UNSPSC Code:
12352106
EC Index Number:
213-988-5
NACRES:
NA.22
form:
powder

Quality Level

form

powder

potency

5820 mg/kg LD50, oral (Rat)

mp

241-243 °C

storage temp.

2-30°C

SMILES string

[P](=O)(OCCN)(O)O

InChI

1S/C2H8NO4P/c3-1-2-7-8(4,5)6/h1-3H2,(H2,4,5,6)

InChI key

SUHOOTKUPISOBE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service