8.01309
4-Amino-1,2,4-triazole
for synthesis
Synonym(s):
4-Amino-1,2,4-triazole, 4-Amino-4H-1,2,4-triazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
MDL number:
UNSPSC Code:
12352116
EC Index Number:
209-533-5
NACRES:
NA.22
Recommended Products
Quality Level
form
powder
pH
<7 (20 °C in H2O)
mp
83-86 °C
storage temp.
2-30°C
SMILES string
[n]1(cnnc1)N
InChI
1S/C2H4N4/c3-6-1-4-5-2-6/h1-2H,3H2
InChI key
FMCUPJKTGNBGEC-UHFFFAOYSA-N
Analysis Note
Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 83 °C
Melting range (upper value): ≤ 86 °C
Identity (IR): passes test
Melting range (lower value): ≥ 83 °C
Melting range (upper value): ≤ 86 °C
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service