539322
Protein G, Peroxidase Conjugate
This Protein G, Peroxidase Conjugate is validated for use in Direct Immunoassay.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116133
NACRES:
NA.54
Recommended Products
Quality Level
form
liquid
contains
0.02% Thimerosal as preservative
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
shipped in
wet ice
storage temp.
2-8°C
General description
Purified recombinant protein G conjugated to peroxidase.
Purified recombinant protein G conjugated to peroxidase. Binds ≥ 99% to human IgG immobilized on a column.
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Analysis Note
Single band at 33.8 kDa by SDS-PAGE
Other Notes
Binds ≥99% to human IgG immobilized on a column. o-Phenylenediamine, 1 mg/ml in citrate buffer, pH 4.5, 0.1% urea hydrogen peroxide at 25°C, is used for testing immunoenzyme reactivity Purity of the recombinant protein G has been determined by presence of a single band at M.W. 33,800 on a 15% SDS-polyacrylamide gel. Specifically binds the Fc region of IgG from various species. Variables associated with assay conditions will dictate the proper working dilution.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gleyder Roman-Sosa et al.
Vaccines, 10(2) (2022-02-27)
New World arenaviruses are rodent-transmitted viruses and include a number of pathogens that are responsible for causing severe human disease. This includes Junín virus (JUNV), which is the causative agent of Argentine hemorrhagic fever. The wild nature and mobility of
M J Weickert et al.
Applied and environmental microbiology, 63(11), 4313-4320 (1997-11-15)
Accumulation of soluble recombinant hemoglobin (rHb1.1) in Escherichia coli requires proper protein folding, prosthetic group (heme) addition, and subunit assembly. This served as a new model system for the study of the effects of temperature, protein synthesis rates, and protein
L M Nemec et al.
Journal of dairy science, 95(8), 4568-4577 (2012-07-24)
The aim of this study was to compare effects of inorganic sulfate versus chelated forms of supplemental Cu, Mn, and Zn on milk production, plasma and milk mineral concentrations, neutrophil activity, and antibody titer response to a model vaccination. Holstein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service