Skip to Content
Merck
All Photos(1)

Key Documents

5.30536

Sigma-Aldrich

EGFR Activator, NSC 228155

Synonym(s):

EGFR Activator, NSC 228155, 4-Nitro-7-((1-oxidopyridin-2-yl)sulfanyl)-2,1,3-benzoxadiazole, 4-Nitro-7-(1-oxy-pyridin-2-ylsulfanyl)-benzo[1,2,5]oxadiazole, 2-(7-Nitrobenzo[c][1,2,5]oxadiazol-4-ylthio)pyridine-1-oxide, NSC-228155

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6N4O4S
CAS Number:
Molecular Weight:
290.25
MDL number:
UNSPSC Code:
41121801
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

burnt orange

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

SMILES string

S(c3[n+](cccc3)[O-])c1c2n[o]nc2c(cc1)[N+](=O)[O-]

InChI key

ICCFXXDUYSPKOL-UHFFFAOYSA-N

General description

A nitro-benzoxadiazole (NBD) compound that directly binds to the dimerization domain of sEGF receptor to generate stable dimers and allosterically activates the receptor. Exhibits similar phosphorylation profile as the cognate EGF-ligand. Enhances the phosphorylation of Tyr1068 and Tyr1173 in MDA-MB-468 cells in a time- and dose-dependent manner (EC50 = 52 µM with respect to Tyr1068). These phosphorylation events can be blocked by pre-incubating cells with AG1478 (>Cat. No. 658852; 10 µM) or PD153035 (https://www.emdmillipore.com/US/en/product/Complement-C9-Depleted-Serum-Human,EMD_BIO-234409">>Cat. No. 234490; 2 µM) that selectively block the ATP-binding pocket of EGFR. Tyr1068 phosphorylation of the receptor can also be completely blocked in EGF treated MDA-MB-468 cells exposed to anti-EGFR neutralizing antibody.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGF receptor

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Sakanyan, V., et al. 2014. Sci. Rep.4, 3977.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vehary Sakanyan et al.
Scientific reports, 4, 3977-3977 (2014-02-06)
Peptide ligand-induced dimerization of the extracellular region of the epidermal growth factor receptor (sEGFR) is central to the signal transduction of many cellular processes. A small molecule microarray screen has been developed to search for non-peptide compounds able to bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service