527624
Plasmin, EACA- and Lysine-Free, Human Plasma
Synonym(s):
Human Plasma Plasmin, Human Plasmin, Lysine-Free Plasmin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Merck USA index - 14, 7520
Quality Level
form
lyophilized
specific activity
≥7 units/mg protein
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
solubility
water: soluble
shipped in
ambient
storage temp.
−20°C
General description
Native, EACA- and lysine-free plasma from human plasma. Catalyzes the hydrolysis of peptides, arginine and lysine esters and converts fibrin into soluble products.
Native, EACA- and lysine-free plasma from human plasma. Catalyzes the hydrolysis of peptides, arginine and lysine esters, and converts fibrin into soluble products.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of N-tosyl-Gly-Pro-Lys-pNA per min at 25°C, pH 7.8.
Physical form
Lyophilized from 20 mM sodium phosphate buffer, 10 mg D-mannitol, 10 mg NaCl.
Preparation Note
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
From Catalog:
Desc. Field- added "Catalyzes the hydrolysis of peptides, arginine and lysine esters and converts fibrin into soluble products."
From Catalog:
Desc. Field- added "Catalyzes the hydrolysis of peptides, arginine and lysine esters and converts fibrin into soluble products."
Other Notes
Federici, A.B., et al. 1993. Blood81, 720.
Hoffmann, J.J., and Janssen, W.C. 1992. Thromb. Res.67, 711.
Hoffmann, J.J., and Janssen, W.C. 1992. Thromb. Res.67, 711.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service