Skip to Content
Merck
All Photos(2)

Key Documents

516567

Sigma-Aldrich

PPARβ/δ Antagonist, GSK3787

The PPARβ/δAntagonist II, PT-S58, also referenced under CAS 188591-46-0, controls the biological activity of PPARβ/δ. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

PPARβ/δ Antagonist, GSK3787, 4-Chloro-N-(2-((5-trifluoromethyl)-2-pyridyl)sulfonyl)ethyl)benzamide, PPARβ Antagonist I, PPARδ Antagonist I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClF3N2O3S
CAS Number:
Molecular Weight:
392.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cnc(cc1)[S](=O)(=O)CCNC(=O)c2ccc(cc2)Cl

InChI

1S/C15H12ClF3N2O3S/c16-12-4-1-10(2-5-12)14(22)20-7-8-25(23,24)13-6-3-11(9-21-13)15(17,18)19/h1-6,9H,7-8H2,(H,20,22)

InChI key

JFUIMTGOQCQTPF-UHFFFAOYSA-N

General description

A cell-permeable and orally available pyridylsulfone compound that acts as a selective, high affinity PPARβ (PPARδ) ligand (IC50 in PPAR ligand displacement assays = 200 nM against human PPARβ and >10 µM against PPARα or PPARγ) and effectively antagonizes agonist-induced, but not basal, PPARβ transcription activity both in cultures in vitro (IC50 = 126 nM against 2 nM GW501516-induced reporter transcription in CV-1 cells) and in mice in vivo (82% and 71% inhibition of 10 mg/kg GW0742-induced Adrp and Angptl4 mRNA upregulation, respectively, in colon epithelium by 10 mg/kg GSK3787; p.o.) by covalently modifying PPARβ at Cys249. GSK3787 is also shown to exhibit weak agonistic activity toward PPARγ (1.2-fold increase above basal level by 1 µM GSK3787), and effectively block the more potent agonist GW1929 (Cat. Nos. 370695) from further stimulation (1.5 and 3.5-fold above basal by 0.3 µM GW1929 in the presence or absence of 1 µM GSK3787, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Palkar, P.S., et al. 2010. Mol. Pharmacol.78, 419.
Shearer, B,G., et al. 2010. J. Med. Chem.53, 1857.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service