Skip to Content
Merck
All Photos(1)

Key Documents

5.09663

Sigma-Aldrich

MCT1 Inhibitor, SR13800

Synonym(s):

MCT1 Inhibitor, SR13800, Monocarboxylate Transport I Inhibitor, SLC16A1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29N3O2S
CAS Number:
Molecular Weight:
435.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyrole pyrimidine-based compound that acts as an inhibitor of monocarboxylate transporter 1 (MCT1, SLC16A1) and blocks the proliferation of Raji Burkitt lymphoma (Raji BL) cells expressing MCT1 (EC50 = 5 nM). Also shown to inhibit the growth of MCF-7 breast cancer cells. Effectively increases intracellular lactate levels while reducing lactate export and intracellular ATP levels in Raji cells. However, it does not affect basal oxygen consumption or the mitochondrial mass in these cells. Delays or even eliminates tumor growth in NOD/SCID mice injected with Raji lymphoma cells. Acts synergistically with metformin (Cat. No. 317240) to block monocarboxylate transport and tumor growth in humans and in murine models. Their combined effect also reduces cellular levels of glutathione with concomitant increase in hydrogen peroxide levels, which increases the death of Raji BL cells.
A cell-permeable pyrole pyrimidine-based compound that acts as an inhibitor of monocarboxylate transporter 1 (MCT1, SLC16A1) and blocks the proliferation of Raji Burkitt lymphoma (Raji BL) cells expressing MCT1 (EC50 = 5 nM). Also shown to inhibit the growth of MCF-7 breast cancer cells. Effectively increases intracellular lactate levels while reducing lactate export and intracellular ATP levels in Raji cells. However, it does not affect basal oxygen consumption or the mitochondrial mass in these cells. Delays or even eliminates tumor growth in NOD/SCID mice injected with Raji lymphoma cells. Acts synergistically with metformin (Cat. No. 317240) to block monocarboxylate transport and tumor growth in humans and in murine models. Their combined effect also reduces cellular levels of glutathione with concomitant increase in hydrogen peroxide levels, which increases the death of Raji BL cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MCT1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Doherty, J., et al. 2014. Cancer Res.73,908.
Murray, C., et al. 2013. Nature. Chem. Biol.1, 371.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service