Skip to Content
Merck
All Photos(1)

Documents

5.09228

Sigma-Aldrich

Y-27632

≥95% (HPLC), liquid, Rho kinase inhibitor, Calbiochem

Synonym(s):

InSolution Y-27632, Sterile-Filtered, Cell Culture-Tested

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O · 2HCl
CAS Number:
Molecular Weight:
320.26
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Y-27632, InSolution, ≥95%, 10 mM aqueous solution, Sterile-Filtered, inhibitor of Rho-associated protein kinases

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−20°C

InChI

1S/C14H21N3O/c1-10(15)11-2-4-12(5-3-11)14(18)17-13-6-8-16-9-7-13/h6-12H,2-5,15H2,1H3,(H,16,17,18)/t10-,11?,12?/m1/s1

InChI key

IYOZTVGMEWJPKR-VOMCLLRMSA-N

General description

A 10 mM (2 mg/591 µL) sterile-filtered solution of Y-27632 in H₂O.
Ready to use, cell culture-tested, suitable for stem cell applications. Acts as a highly potent, cell-permeable, reversible, and selective inhibitor of Rho-associated protein kinases (Ki = 140 nM for p160ROCK). Also shown to improve survival of human stem cells especially during cell culture passage and manipulation.

Application

InSolution Y-27632, Sterile-Filtered, Cell Culture-Tested, CAS 146986-50-7, is a 10 mM sterile-filtered solution of Y-27632 in H₂O. Acts as an inhibitor of Rho kinases (Ki = 140 nM for p160ROCK).

Biochem/physiol Actions

Primary Target
ROCK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 10 mM (2 mg/591 µL) sterile-filtered solution of Y-27632 (Cat. No. 688000) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Li X. et al, et. Al., 2009. Human Reproduction, 24, 580.
Chitaley, K., et al. 2001. Nat. Med.7, 119.
Davies, S.P., et al. 2000. Biochem. J.351, 95.
Narumiya, S., et al. 2000. Methods Enzymol.325, 273.
Hirose, M., et al. 1999. J. Cell Biol.141, 1625.
Maekawa, M., et al. 1999. Science.285, 895.
Uehata, M., et al. 1997. Nature.389, 990.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service