Skip to Content
Merck
All Photos(1)

Key Documents

5.04045

Sigma-Aldrich

Focal Adhesion Kinase Inhibitor III

Synonym(s):

Focal Adhesion Kinase Inhibitor III, N-(4- tert-Butylbenzyl)-1,5-dimethyl-1,5-dihydropyrazolo[4,3-c][2,1]benzothiazin-8-amine-4,4-dioxide, FAK Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N4O2S
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable pyrazolobenzothiazinedioxide compound that targets and locks FAK in its inactive conformation, resulting in preincubation time-dependent inhibition in kinase assays and FAK autophosphorylation in human prostate cancer PC3M-luc cells (IC50 = 7.1 µM with 30 min inhibitor preincubation), while exhibiting little activity toward Akt1, Aurora B, HER2, MEK1, p38α, Pyk2, or Src even at concentrations as high as 10 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FAK

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tomita, N., et al. 2013. Bioorg. Med. Chem. Lett.23, 1779.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service