Skip to Content
Merck

5.00949

SAFC

all-rac-α-Tocopherol

EMPROVE® ESSENTIAL Ph Eur,BP,USP,E 307

Pharma Manufacturing

Synonym(s):

all-rac-α-Tocopherol, Vitamin E alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
MDL number:
UNSPSC Code:
12352104
E Number:
E307
EC Index Number:
233-466-0
Pricing and availability is not currently available.

Agency

BP
Ph. Eur.
USP

Quality Level

vapor pressure

<1 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

viscous liquid

autoignition temp.

280 °C

potency

>4000 mg/kg LD50, oral (Rat)

bp

393 °C/1013 hPa

mp

2.5-3.5 °C

density

0.95 g/cm3 at 20 °C

application(s)

pharmaceutical

storage temp.

15-25°C

SMILES string

O1[C@@](CCc2c1c(c(c(c2C)O)C)C)(CCC[C@@H](CCC[C@@H](CCCC(C)C)C)C)C

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1

InChI key

GVJHHUAWPYXKBD-IEOSBIPESA-N

General description

As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

all-rac-α-Tocopherol (Vitamin E) protects cells against free radicals. Vitamin E is used in skin and hair care products, as well as in dietary supplements. all-rac-α-Tocopherol Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service