Skip to Content
Merck
All Photos(1)

Key Documents

481913

Millipore

NADH, Disodium Salt

Functions as a coenzyme of a large number of oxidoreductases.

Synonym(s):

NADH, Disodium Salt, β-Nicotinamide-adenine Dinucleotide, Reduced, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · 2Na
CAS Number:
Molecular Weight:
709.40
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

Quality Level

description

Merck USA index - 14, 6344

Assay

≥95% dry basis (enzymatic)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

yellow

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])(O[P](=O)(OC[C@H]4O[C@H]([C@@H]([C@@H]4O)O)[n+]5cc(ccc5)\C(=N\[H])\[O-])O)OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n]2c3ncnc(c3nc2)N

InChI

1S/C21H27N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37);;/q;2*+1/p-1/t10-,11-,13-,14-,15-,16-,20-,21-;;/m1../s1

InChI key

GUIOTHWMFMEYPG-WUEGHLCSSA-M

General description

Functions as a coenzyme of a large number of oxidoreductases.

Warning

Toxicity: Standard Handling (A)

Other Notes

During handling of the material, it is very important that the container is placed at room temperature for a sufficient amount of time to allow the material to reach room temperature before the container is opened

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service