Skip to Content
Merck

4.80791

SAFC

Hydrochloric acid

5 mol/l, EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Hydrochloric acid, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
12352106
EC Index Number:
231-595-7

grade

GMO free
GMP
Halal
Kosher

Quality Level

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

product line

EMPROVE® EXPERT

bp

>100 °C (lit.)

density

1.18 g/L at 20 °C

application(s)

cleaning in place
pharma/biopharma processes

foreign activity

Aflatoxin

storage temp.

2-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Packaging

4.80791.5000 / 5l / PE bottle
4.80791.9203/ 200l / PE drum

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service