Skip to Content
Merck
All Photos(1)

Key Documents

474502

Sigma-Aldrich

4-Methylumbelliferyl-N-acetyl-β-D-glucosaminide

Ultrasensitive, fluorogenic substrate for assays of N-acetyl-β-D-glucosaminidase.

Synonym(s):

4-Methylumbelliferyl-N-acetyl-β-D-glucosaminide, 4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO8
CAS Number:
Molecular Weight:
379.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

solubility

methanol: soluble
water: soluble

fluorescence

λex 360 nm
λem 440 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@@H]1[C@H]([C@@H]([C@H](OC1Oc2cc3c(cc2)C(=CC(=O)O3)C)CO)O)O)C(=O)C

InChI

1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16-,17-,18?/m1/s1

InChI key

QCTHLCFVVACBSA-XWEABGILSA-N

General description

Ultrasensitive, fluorogenic substrate for assays of N-acetyl-β-D-glucosaminidase (excitation maximum: 360 nm; emission maximum: 440 nm).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in aqueous solutions. Reconstitute just prior to use.
Unstable in solution; reconstitute just prior to use.

Other Notes

Chen, K.S., et al. 1994. Electrophoresis 15, 662.
Leaback, D.H., and Walker, P.G. 1961. Biochem. J.78, 151.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service