Skip to Content
Merck
All Photos(1)

Key Documents

361551

Sigma-Aldrich

GSK-3 Inhibitor X

The GSK-3 Inhibitor X, also referenced under CAS 740841-15-0, controls the biological activity of GSK-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3 Inhibitor X, (2ʹZ,3ʹE)-6-Bromoindirubin-3ʹ-acetoxime, BIO-Acetoxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12BrN3O3
CAS Number:
Molecular Weight:
398.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark purple

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

An acetoxime analog of BIO, GSK-3 Inhibitor IX (Cat. No. 361550) that exhibits greater selectivity for GSK-3α/β (IC50 = 10 nM) over Cdk5/p25, Cdk2/A and Cdk1/B (IC50 = 2.4 µM, 4.3 µM and 63 µM, respectively). Weakly affects the activities of Cdk4/D1 and many other kinases (IC50 ≥ 10 µM). Also acts as an aryl hydrocarbon receptor agonist in both yeast (EC50 ≥ 0.1 µM) and mammalian (EC50 = 0.16 µM) reporter systems.

Biochem/physiol Actions

Cell permeable: no
Primary Target
GSK-3α/β
Product does not compete with ATP.
Reversible: no
Target IC50: 10 nM against GSK-3α/β

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Preparation Note

Limited solubility in aqueous buffers; serial dilutions in DMSO may be required prior to dilution in buffer.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Knockaert, M., et al. 2004. Oncogene23, 4400.
Polychronopoulos, P., et al. 2004. J. Med. Chem.47, 935.
Meijer, L., et al. 2003. Chem. Biol.10, 1255.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noboru Sato et al.
Methods in molecular biology (Clifton, N.J.), 331, 115-128 (2006-08-03)
This chapter introduces a new method of maintaining human embryonic stem cells (hESCs) in the undifferentiated state through treatment with a GSK-3 inhibitor, BIO, under a feeder-free condition. Additionally, methods are introduced that determine multidifferentiation potential of hESCs by differentiating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service