Skip to Content
Merck
All Photos(1)

Documents

3097S

Sigma-Aldrich

NucleoClean Decontamination Spray

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

form

liquid

Quality Level

manufacturer/tradename

Chemicon®

shipped in

ambient

General description

NucleoClean<TMSYMBOL></TMSYMBOL> Decontamination spray is the simple solution for DNA Decontamination.
Millipore′s NucleoClean<TMSYMBOL></TMSYMBOL> Decontamination spray will keep your lab free of PCR carryover contamination. Our NucleoClean<TMSYMBOL></TMSYMBOL> spray offers you a simple and effective way to clean your PCR equipment, labware, and benches --a quick, easy, and safe alternative to bleaching methods!

Stable at room temperature for months when properly closed.

Millipore also offers the AMP ID system, a highly efficient nucleic acid amplification technology configured in a convenient single tube reaction format for Real-Time PCR detection of RNA or DNA based targets. Each kit supplies enough reagent for as many as 100 assays and includes optimized positive controls and amplification primers specifically designed for the detection of select infectious agents. The AMP ID line currently includes kits for Pan-Enterovirus (AMP3100), West Nile Virus (AMP3200), Avian Influenza A Virus, H5N1 (AMP3500), and Herpes Simplex Virus Type 1 and Type 2 (AMP3800). In addition Millipore offers ViralXpress, a powerful nucleic acid extraction and purification system, specifically intended for the highly efficient purification of viral DNA and RNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, infected cells or cell culture supernatants (3095). Each kit supplies enough reagents for up to 50 extractions.

Application

NucleoClean Decontamination Spray is suitable for the decontamination of nucleic acids on labware, equipment and lab benches.

Linkage

Replaces: 3097

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service