239819
CXCR2 antagonist, Cpd 19
The CXCR2 antagonist, Cpd 19, also referenced under CAS 473728-67-5, controls the biological activity of CXCR2.
Synonym(s):
CXCR2 antagonist, Cpd 19, 3-(2-(cyclopentylamino)-3,4-dioxocyclobut-1-enylamino)-2-hydroxy-N,N-dimethylbenzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H21N3O4
CAS Number:
Molecular Weight:
343.38
UNSPSC Code:
12352200
NACRES:
NA.51
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 50 mg/mL, light brown
shipped in
ambient
storage temp.
−20°C
General description
A cell-permeable, cyclobutenedione derivative that acts as a potent antagonist of chemokine receptor CXCR2 (IL8RB) (IC50 = 8 nM). Reported to inhibit CXCR2-mediated chemotaxis in a CXCR2 expressing cell line (IC50 = 145 nM). Exhibits good stability in human and rat liver microsomal preparations (>50% remaining after 30 min at 37°C).
Packaging
Packaged under inert gas
Warning
Toxicity: Regulatory Review (Z)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Merritt, J., et al. 2006. Bioorg. Med. Chem. Lett.16, 4107.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service