Skip to Content
Merck
All Photos(1)

Key Documents

176901

Sigma-Aldrich

Anthrax Lethal Factor Protease Inhibitor, In-2-LF

The Anthrax Lethal Factor Protease Inhibitor, In-2-LF controls the biological activity of Anthrax Lethal Factor Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Anthrax Lethal Factor Protease Inhibitor, In-2-LF, Anthrax LF Protease Inhibitor, Bacillus anthracis LF Protease Inhibitor, In-2-LF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C81H150N42O17
Molecular Weight:
1984.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL
5% acetic acid: 5 mg/mL
DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable N-acetylated, C-hydroxamate derivative of a 14-mer peptide designed from the MEK-2 template that acts as a competitive inhibitor of Anthrax lethal factor (LF) metalloprotease (Ki = 1 nM). Also inhibits MEK-3 cleavage. Protects against Anthrax toxin induced cytotoxicity in RAW264.7 and J772.A1 cells.
A cell-permeable derivative of a 14-mer peptide that is based on the MEK-2 template and acts as a competitive inhibitor of Anthrax lethal factor metalloprotease (Ki = 1 nM). Also inhibits MEK-3 cleavage. Exhibits protective effects against Anthrax toxin-induced cytotoxicity in RAW264.7 and J772.A1 cells. Reported to competitively inhibit furin (Ki = 49 nM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
anthrax LF metalloprotease
Product does not compete with ATP.
Reversible: no
Target Ki: 1 nM against Anthrax lethal factor (LF) metalloprotease

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Gly-Tyr-βAla-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Val-Leu-Arg-NHOH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Peinado, J.R., et al. 2004. Biochem. Biophys. Res. Commun.321, 601.
Tonello, F., et. al. 2002. Nature418, 386.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service