171272
AMPK Activator VI, RSVA314
The AMPK Activator VI, RSVA314 modulates the biological activity of AMPK. This small molecule/inhibitor is primarily used for Biochemicals applications.
Synonym(s):
AMPK Activator VI, RSVA314, RSVA314, AMPK Signaling Activator VI
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H21N3O3
CAS Number:
Molecular Weight:
327.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51
Recommended Products
Quality Level
Assay
>90% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
yellow
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
A cell permeable hydrozide that activates AMPK indirectly via a CaMKKβ-dependent pathway similar to resveratrol (Cat. No. 554325), which results in the phosphorylation of Thr-172 on the AMPKα2 subunit. This activator promotes lysosomal amyloid-β degradation in 3 cell lines nearly 40 times more potently than resveratrol with an IC50 of 1-2 µM. Treatment with RSVA314 at 1-3 µM results in a dose-dependent inhibition of the phosphorylation of 3 proteins downstream from mTOR activation including p70S6K, eukaryotic elongation factor 2 kinase (eEF2K), and S6 ribosomal protein, and promotes a robust increase in the phosphorylation of the protein raptor at Ser-792 in western blots. At the same concentrations, it elicits a strong, dose-dependent conversion of the light chain 3 (LC3) protein, a marker involved with autophagy induction, from LC3-I and LC3-II, also in western blots.
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Other Notes
Vingtdeux, V., et al. 2011. FASEB J.25, 219.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service