Skip to Content
Merck

1.37120

SAFC

Benzyl alcohol

EMPROVE® EXPERT, Ph. Eur., ChP, JP, NF

Pharma Manufacturing

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
51241802
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Agency

ChP
JP
NF
Ph. Eur.

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

product line

EMPROVE® EXPERT

Assay

≥98% (GC)
≥98.0-100.5% (titration)

form

liquid

autoignition temp.

817 °F

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Benzyl alcohol is typically used as a preservative in liquid formulations and is active against bacteria, yeast and mould. It can also serve as a solvent for poorly soluble drugs.

Benzyl Alcohol Emprove® Expert is an extraordinarily pure product with impurity specification thresholds even below current requirements of designated pharmacopoeias (e.g. benzaldehyde content < 0.05 %). Therefore, this product is especially suitable for high-risk applications or any other purpose that requires superior quality.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

Find key considerations for the selection and use of benzalkonium chloride, benzyl alcohol, benzaldehyde, parabens, benzoic acid, and sodium benzoate as preservatives.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service