Skip to Content
Merck

1.37074

SAFC

2-Morpholinoethanesulfonic acid monohydrate (MES)

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

2-Morpholinoethanesulfonic acid monohydrate (MES), MES

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
213.25
MDL number:
UNSPSC Code:
12161703
EC Index Number:
224-632-3

Quality Level

product line

EMPROVE® EXPERT

feature

RNase and DNase free

pH

3.6 (20 °C, 10 g/L in H2O)

mp

>300 °C

solubility

80 g/L

bulk density

570 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)CCN1CCOCC1

InChI

1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)

InChI key

SXGZJKUKBWWHRA-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.
Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service