1.09971
Hydrochloric acid solution
c(HCl) = 0.5 mol/l (0.5 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®
Synonym(s):
Hydrochloric acid solution, Hydrogen chloride solution
About This Item
Recommended Products
Product Name
Hydrochloric acid solution, for 1000 ml, c(HCl) = 0.5 mol/l (0.5 N) Titrisol®
Quality Level
product line
Titrisol®
form
liquid
reaction suitability
reaction type: Acid-base reactions
concentration
0.5 M
technique(s)
titration: suitable
pH
<1 (20 °C in H2O)
density
1.1 g/cm3 at 20 °C
storage temp.
15-25°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc.: This study explored the utilization of galvanic waste sludge as a zinc source in synthesizing zinc-ammonium phosphate. Hydrochloric acid solution played a crucial role in the extraction process of zinc from the waste sludge, showcasing its application in the efficient recovery of valuable metals from industrial waste (Morgovan et al., 2024).
- Contactless conductivity sensor employing moist paper as absorbent for in-situ detection of generated carbon dioxide gas.: This paper presents a novel analytical method using a contactless conductivity sensor with moist paper as an absorbent for detecting CO2. Hydrochloric acid solution was utilized in the calibration and preparation of the sensing materials, emphasizing its importance in developing advanced analytical techniques (Sonsa-Ard et al., 2020).
- sym-Trisubstituted 1,3,5-Triazine Derivatives as Promising Organic Corrosion Inhibitors for Steel in Acidic Solution.: This research investigates the effectiveness of trisubstituted triazine derivatives as corrosion inhibitors for steel in hydrochloric acid solution. The study highlights the solution′s critical role in simulating industrial conditions and evaluating the inhibitors′ performance (El-Faham et al., 2016).
- Effect of molecular structure of aniline-formaldehyde copolymers on corrosion inhibition of mild steel in hydrochloric acid solution.: The study examines how the molecular structure of aniline-formaldehyde copolymers affects their ability to inhibit corrosion in mild steel. Hydrochloric acid solution was used to create the corrosive environment, underscoring its application in corrosion studies and material testing (Zhang et al., 2015).
- Graphene sheets modified with polyindole for electro-chemical detection of dopamine.: This paper discusses the modification of graphene sheets with polyindole to enhance the electrochemical detection of dopamine. Hydrochloric acid solution was employed in the preparation of the modified graphene sheets, demonstrating its role in nanomaterial fabrication for analytical chemistry applications (Kumar and Prakash, 2014).
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service