Skip to Content
Merck
All Photos(1)

Key Documents

1.06513

SAFC

Sodium thiosulfate pentahydrate

pure

Synonym(s):

Sodium thiosulfate pentahydrate, Antichlor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O3S2 · 5H2O
CAS Number:
Molecular Weight:
248.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-867-5

Quality Level

form

solid

pH

6.0-7.5 (20 °C, 100 g/L in H2O)

mp

48 °C

solubility

701 g/L

density

1.74 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=S)(=O)([O-])[O-].O.O.O.O.O

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Application

Sodium thiosulfate pentahydrate is used as antioxidant or chelating agent and is suitable for use in pharmaceutical processes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service