Skip to Content
Merck
All Photos(1)

Key Documents

1.06067

Supelco

Magnesium sulfate

anhydrous for analysis EMSURE®

Synonym(s):

Magnesium sulfate anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO4S
CAS Number:
Molecular Weight:
120.37
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-298-2
NACRES:
NA.21
Assay:
≥98.0% (complexometric)
grade:
anhydrous
form:
solid

grade

anhydrous

Quality Level

Agency

suitable for SM 5210

product line

EMSURE®

Assay

≥98.0% (complexometric)

form

solid

impurities

≤0.004% Total nitrogen (N)

loss

≤2% loss on ignition, 600 °C

pH

7.9 (25 °C, 50 g/L in H2O)

mp

1124 °C

density

2.66 g/cm3 at 20 °C

bulk density

600 kg/m3

anion traces

chloride (Cl-): ≤0.001%

cation traces

As: ≤0.0001%
Ca: ≤0.04%
Fe: ≤0.001%
Mn: ≤0.002%
N: ≤0.004%
heavy metals (as Pb): ≤0.001%

storage temp.

no temp limit

SMILES string

[Mg+2].[S](=O)(=O)([O-])[O-]

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Application

  • Bioflocculant produced by Bacillus velezensis and its potential application in brewery wastewater treatment.: This study explores the production of a bioflocculant by Bacillus velezensis, assessing its application in treating brewery wastewater. Magnesium sulfate was used as a coagulant aid, significantly improving the flocculation process (Agunbiade et al., 2022).
  • Evaluation of the automated micro-solid phase extraction clean-up system for the analysis of pesticide residues in cereals by gas chromatography-Orbitrap mass spectrometry.: This research evaluates an automated micro-solid phase extraction system for analyzing pesticide residues. Magnesium sulfate was utilized for sample clean-up, enhancing the extraction efficiency and accuracy (Hakme and Poulsen, 2021).
  • Online supercritical fluid extraction and chromatography of biomarkers analysis in aqueous samples for in situ planetary applications.: This paper discusses a method combining supercritical fluid extraction and chromatography for biomarker analysis. Magnesium sulfate played a crucial role in the extraction process, facilitating the separation of target compounds (Abrahamsson et al., 2019).
  • Bioremediation of oily hypersaline soil and water via potassium and magnesium amendment.: The study examines the bioremediation of oily hypersaline environments using magnesium sulfate and potassium. The amendments improved microbial activity and degradation of hydrocarbons (Al-Mailem et al., 2013).

Analysis Note

Assay (complexometric): ≥ 98.0 %
Chloride (Cl): ≤ 0.001 %
Total nitrogen (N): ≤ 0.004 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.04 %
Fe (Iron): ≤ 0.001 %
Mn (Manganese): ≤ 0.002 %
Loss on ignition (600 °C): ≤ 2 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service