Skip to Content
Merck
All Photos(1)

Documents

1.04159

Sigma-Aldrich

Orcein

Sign Into View Organizational & Contract Pricing


About This Item

NACRES:
NA.21

form

solid

Quality Level

bulk density

900‑1000 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

Related Categories

General description

The dry dye Orcein for microscopy Certistain® is used for the preparation of a staining solution for staining of elastic fibers. It is also used for sex chromatin staining, nuclear staining and demonstration of hepatic inclusions, especially of hepatitis B antigen. As a CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (NaOH 0.01 mol/l): 575 - 580 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.001 %; Sodium hydroxide solution 0.01 mol/l): 270 - 330
TLC-Test: passes test
Loss on drying (110 °C): ≤ 7 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service