1.02122
LICHROLUT® RP-18 solid phase extraction (SPE) tubes
(40 - 63 µm) 1000mg 6ml, package of 30 tubes
About This Item
Recommended Products
Product Name
LiChrolut® RP-18 (40 - 63 µm) 1000 mg 6 ml standard PP-tubes, package of 30 tubes
packaging
package of 30 tubes
technique(s)
solid phase extraction (SPE): suitable
storage temp.
no temp limit
Related Categories
General description
Analysis Note
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Explore our selection of high-quality LiChrolut® SPE products for efficient and reliable sample preparation before chromatographic analysis.
Explore our selection of high-quality LiChrolut® SPE products for efficient and reliable sample preparation before chromatographic analysis.
Explore our selection of high-quality LiChrolut® SPE products for efficient and reliable sample preparation before chromatographic analysis.
Explore our selection of high-quality LiChrolut® SPE products for efficient and reliable sample preparation before chromatographic analysis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service