Skip to Content
Merck

1.01700

SAFC

L-Alanine

EMPROVE® EXPERT, Ph. Eur., JP, USP

Pharma Manufacturing

Synonym(s):

L-Alanine, 2-Aminopropionic acid, Ala

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
89.09
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-273-8
Pricing and availability is not currently available.

Agency

JP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® EXPERT

pH

5.5-6.5 (20 °C, 100 g/L in H2O)

mp

295-297 °C (decomposition)

solubility

166.5 g/L

density

1.40 g/cm3 at 20 °C

bulk density

660 kg/m3

application(s)

parenterals ()

storage temp.

15-25°C

SMILES string

[N+H3]C(C)C(=O)[O-]

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)

InChI key

QNAYBMKLOCPYGJ-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service