Recommended Products
Quality Level
form
liquid
IVD
for in vitro diagnostic use
bp
100 °C
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
General description
The ready-to-use Picrofuchsin solution acc. to van Gieson for Microscopy, is used for staining collagenous connective tissue, muscle tissue, and cornified epithelium in human sample material. It can also be used to stain glia fibrils and cytoplasm. Fuchsin and picric acid, the two ingredients of the solution, stain the various tissue structures simultaneously and selectively. Collagenous fibers will appear with a vibrant red color, muscle tissue and glia fibrils will be stained yellow and amyloid, hyaline, colloid, as well as mucus will be stained gradually red to yellow. Staining with Weigert′s iron hematoxylin is advised to produce a stable and persistent stain of the nuclei. Picrofuchsin solution acc. to van Gieson can also be used in combination with elastin solution acc. to Weigert in the van Gieson elastica staining method to detect elastic fibers in histological sections. 500 ml of the Pirofuchsin solution are sufficient for at least 500 specimens. It is an IVD and CE registered product and can be applied for diagnostical purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Application
For staining collagenous connective tissue, muscle tissue, and cornified epithelium
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service