Skip to Content
Merck
All Photos(1)

Documents

07-532

Sigma-Aldrich

Anti-phospho-PKR (Thr446) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr446)

Gene Information

human ... EIF2AK2(5610)

Related Categories

Specificity

phospho-PKR

Immunogen

Peptide corresponding to amino acids surrounding Thr446 of human PKR.

Application

Anti-phospho-PKR (Thr446) Antibody is an antibody against phospho-PKR (Thr446) for use in WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot in RIPA lysates from HeLa cells that had been stimulated with IFN alpha overnight followed by treatment with calyculin A for 15 minutes

Target description

68 kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Saelens et al.
The Journal of biological chemistry, 276(45), 41620-41628 (2001-09-14)
The protein kinase PKR is a major player in the cellular antiviral response, acting mainly by phosphorylation of the alpha-subunit of the eukaryotic translation initiation factor 2 (eIF2-alpha) to block de novo protein synthesis. PKR activation requires binding of double-stranded
Ka-Chun Suen et al.
The Journal of biological chemistry, 278(50), 49819-49827 (2003-09-17)
One of the hallmarks of Alzheimer's disease is extracellular accumulation of senile plaques composed primarily of aggregated beta-amyloid (Abeta) peptide. Treatment of cultured neurons with Abeta peptide induces neuronal death in which apoptosis is suggested to be one of the
Dario Paolo et al.
Plants (Basel, Switzerland), 10(2) (2021-01-28)
Seed size is the result of complex molecular networks controlling the development of the seed coat (of maternal origin) and the two fertilization products, the embryo and the endosperm. In this study we characterized the role of Arabidopsis thaliana MADS-domain
Shien Hu et al.
Gastroenterology, 133(6), 1893-1904 (2007-12-07)
Inducible heat shock proteins (iHsp), Hsp25/27 and Hsp70, play essential roles in protecting cells against stress and, in intestinal mucosal inflammation, potentially lessening the extent and severity of injury. We examined the expression and regulation of iHsp in human and
Shuen-Ing Tschen et al.
Diabetes, 58(6), 1312-1320 (2009-02-21)
The aim of this study was to elucidate whether age plays a role in the expansion or regeneration of beta-cell mass. We analyzed the capacity of beta-cell expansion in 1.5- and 8-month-old mice in response to a high-fat diet, after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service