Skip to Content
Merck
All Photos(1)

Key Documents

S-051

Supelco

JWH-210 5-Hydroxypentyl metabolite solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27NO2
Molecular Weight:
385.50
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CN(CCCCCO)C2=C1C=CC=C2)C3=CC=C(CC)C4=C3C=CC=C4

InChI

1S/C26H27NO2/c1-2-19-14-15-23(21-11-5-4-10-20(19)21)26(29)24-18-27(16-8-3-9-17-28)25-13-7-6-12-22(24)25/h4-7,10-15,18,28H,2-3,8-9,16-17H2,1H3

InChI key

YKJSXHMELGMWOB-UHFFFAOYSA-N

General description

A Certified Spiking Solution® of JWH-210 5-Hydroxypentyl metabolite. This monohydroxy analog is often referred to as the terminal omega hydroxy metabolite of JWH-210, a synthetic cannabinoid found in several blends of the herbal mixture Spice or K2. The biomarker is a major urinary metabolite of JWH-210.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service