Skip to Content
Merck
All Photos(1)

Key Documents

860527P

Avanti

C24 Ceramide-1-Phosphate (d18:1/24:0)

Avanti Research - A Croda Brand 860527P, powder

Synonym(s):

N-lignoceroyl-ceramide-1-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H87N2O6P
CAS Number:
Molecular Weight:
747.12
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860527P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860527P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI key

JGGZHGYBRJNAOW-FHPBFELKSA-N

General description

Ceramide 1-phosphate (C1P) is a sphingolipid analog of phosphatidate.

Application

C24 Ceramide-1-Phosphate (d18:1/24:0) or N-lignoceroyl-ceramide-1-phosphate (ammonium salt) has been used as a standard to quantify ceramide content in muscle tissue samples using high-performance liquid chromatography/tandem mass spectrometry (LC/MS2).

Biochem/physiol Actions

Ceramide 1-phosphate (C1P) is a mitogenic agent, which has antiapoptotic and prosurvival activities. It negatively regulates tumor necrosis factor-α (TNFα) production stimulated by lipopolysaccharide. C1P also mediates inflammatory response and aids in phagocytosis. It is considered as a second lipid messenger of agonist-stimulated sphingomyelin metabolism.

Packaging

5 mL Amber Glass Screw Cap Vial (860527P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Szczepan Józefowski et al.
Journal of immunology (Baltimore, Md. : 1950), 185(11), 6960-6973 (2010-11-03)
LPS is a constituent of cell walls of Gram-negative bacteria that, acting through the CD14/TLR4 receptor complex, causes strong proinflammatory activation of macrophages. In murine peritoneal macrophages and J774 cells, LPS at 1-2 ng/ml induced maximal TNF-α and MIP-2 release
Lide Arana et al.
Lipids in health and disease, 9, 15-15 (2010-02-09)
Sphingolipids are essential components of cell membranes, and many of them regulate vital cell functions. In particular, ceramide plays crucial roles in cell signaling processes. Two major actions of ceramides are the promotion of cell cycle arrest and the induction
Antonio Gómez-Muñoz et al.
Journal of lipid research, 45(1), 99-105 (2003-10-03)
It was reported previously that ceramide-1-phosphate (Cer-1-P) is mitogenic for fibroblasts (Gómez-Muñoz, A., P. A. Duffy, A. Martin, L. O'Brien, H-S. Byun, R. Bittman, and D. N. Brindley. 1995. Mol. Pharmacol. 47: 883-889; Gómez-Muñoz, A., L. M. Frago, L. Alvarez
A Gomez-Muñoz et al.
Molecular pharmacology, 47(5), 833-839 (1995-05-01)
Ceramide and ceramide-1-phosphate are sphingolipid analogues of diacylglycerol and phosphatidate, respectively, and they are putative second messengers of agonist-stimulated sphingomyelin metabolism. The interactions of exogenous cell-permeable ceramides and ceramide-1-phosphates in modifying DNA synthesis and signal transduction were investigated in Rat-1
V T Hinkovska-Galcheva et al.
The Journal of biological chemistry, 273(50), 33203-33209 (1998-12-05)
Ceramide, a product of agonist-stimulated sphingomyelinase activation, is known to be generated during the phagocytosis of antibody-coated erythrocytes by polymorphonuclear leukocytes. Agonist-stimulated formation of ceramide-1-phosphate is now shown to occur in 32PO4-labeled neutrophils. Ceramide-1-phosphate is formed by a calcium-dependent ceramide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service