Skip to Content
Merck
All Photos(2)

Key Documents

860392P

Avanti

16:0-16:0-d31 PC

Avanti Research - A Croda Brand 860392P, powder

Synonym(s):

1-palmitoyl-2-palmitoyl-d31-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H49NO8PD31
CAS Number:
Molecular Weight:
765.23
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860392P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860392P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(CCCCCCCCCCC

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-16:0-d31 PC has been used to study the phase behavior and organization of sphingomyelin/cholesterol membranes using nuclear magnetic resonance (NMR) spectra.

Packaging

5 mL Amber Glass Screw Cap Vial (860392P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The phase behavior and organization of sphingomyelin/cholesterol membranes: a deuterium NMR study
Keyvanloo A, et al.
Biophysical Journal, 114(6), 1344-1356 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service