Skip to Content
Merck
All Photos(2)

Key Documents

857324P

Avanti

17:0 Cyclic LPA

Avanti Research - A Croda Brand 857324P, powder

Synonym(s):

1-heptadecanoyl-glycero-2,3-cyclic-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H42NO6P
CAS Number:
Molecular Weight:
423.52
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857324P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857324P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P1([O-])O[C@H](COC(CCCCCCCCCCCCCCCC)=O)CO1.[NH4+]

InChI

1S/C20H39O6P.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20(21)24-17-19-18-25-27(22,23)26-19;/h19H,2-18H2,1H3,(H,22,23);1H3/t19-;/m1./s1

InChI key

QQOXNBRKDUFLHW-FSRHSHDFSA-N

General description

Cyclic phosphatidic acid (cPA) is a naturally occurring analog of the growth factor-like phospholipid mediator, lysophosphatidic acid (LPA). The sn-2 hydroxy group of CPA forms a 5-membered ring with the sn-3 phosphate.

Application

17:0 Cyclic LPA or 1-heptadecanoyl-glycero-2,3-cyclic-phosphate (ammonium salt) has been used as an internal standard for the quantification of cyclic lysophosphatidic acid (LPA) esters in proton samples using reversed-phase ultra-performance liquid chromatography-tandem mass spectrometer (UPLC-MS/MS).

Biochem/physiol Actions

cPA affects numerous cellular functions, including anti-mitogenic regulation of the cell cycle, induction of stress fiber formation, inhibition of tumor cell invasion and metastasis, and regulation of differentiation and survival of neuronal cells.
Interestingly, many of these cellular responses caused by cPA oppose those of LPA despite the activation of apparently overlapping receptor populations.

Packaging

5 mL Amber Glass Screw Cap Vial (857324P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Standardization and Quantification of Lysophosphatidic Acid Compounds by Normal-Phase and Reversed-Phase Chromatography-Tandem Mass Spectrometry
Moore JD, et al.
Lysophospholipid Receptors: Signaling and Biochemistry, 137-137 (2013)
Standardization and Quantification of Lysophosphatidic Acid Compounds by Normal-Phase and Reversed-Phase Chromatography-Tandem Mass Spectrometry
Moore JD, et al.
Lysophospholipid Receptors: Signaling and Biochemistry, 137-137 (2013)
Standardization and Quantification of Lysophosphatidic Acid Compounds by Normal-Phase and Reversed-Phase Chromatography-Tandem Mass Spectrometry
Moore JD, et al.
Lysophospholipid Receptors: Signaling and Biochemistry, 137-137 (2013)
Cyclic phosphatidic acid elicits neurotrophin-like actions in embryonic hippocampal neurons.
Fujiwara Y, et al.
Journal of Neurochemistry, 87, 1272-1283 (2003)
Inhibition of cell proliferation by a unique lysophosphatidic acid, PHYLPA, isolated from Physarum polycephalum: signaling events of antiproliferative action by PHYLPA.
Murakami-Murofushi K, et al.
Cell Structure and Function, 18, 363-370 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service