Skip to Content
Merck
All Photos(1)

Key Documents

857144C

Avanti

18:1 BMP-d5 (2R, 2′R/S)

Synonym(s):

sn-(1-oleoyl-2-hydroxy)-glycerol-3-phospho-rac-3′-(1′-oleoyl-2′-hydroxy)-glycerol(d5) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H77D5NO10P
CAS Number:
Molecular Weight:
797.11
UNSPSC Code:
12352211

biological source

synthetic

Assay

≥99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mg (857144C-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NH4+].[2H]C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C([2H])(O)C([2H])([2H])OP([O-])(=O)OC[C@H](O)COC(=O)CCCCCCC\C=C/CCCCCCCC

Application

Lipids research

Packaging

5 mL Amber Glass Screw Cap Vial (857144C-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service