Skip to Content
Merck
All Photos(3)

Documents

840042C

Avanti

Liver PI

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylinositol (Liver, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840042C-10mg)
pkg of 1 × 2.5 mL (840042C-25mg)
pkg of 1 × 4 mL (840042C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840042C-10mg)
10 mg/mL (840042C-25mg)
25 mg/mL (840042C-100mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](O)[C@H](O)[C@@H](O)[C@@H]1O)OP([O-])(OC[C@@]([H])(COC(CCCCCCCCCCCCCCCCC)=O)OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)=O.[Na+]

InChI

1S/C47H83O13P.Na/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(49)59-39(38-58-61(55,56)60-47-45(53)43(51)42(50)44(52)46(47)54)37-57-40(48)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2;/h11,13,17,19,22,24,28,30,39,42-47,50-54H,3-10,12,14-16,18,20-21,23,25-27,29,31-38H2,1-2H3,(H,55,56);/q;+1/p-1/b13-11-,19-17-,24-22-,30-28-;/t39-,42?,43-,44?,45?,46?,47-;/m1./s1

InChI key

NHSOICFDOXKHEC-ZGISKSNKSA-M

General description

Liver PI is a natural phosphatidylinositol. Phosphatidylinositol (PI) is a phosphoglyceride, that has an inositol head group. It is a vital phospholipid component of the plasma membrane.

Application

Liver PI has been used for the preparation of liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (840042C-100mg)
5 mL Clear Glass Sealed Ampule (840042C-10mg)
5 mL Clear Glass Sealed Ampule (840042C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Mechanisms Of Drug Action (2000)
Determination of VPS34/PIK3C3 Activity in vitro Utilising 32P-gammaATP
Munson MJ and Ganley IG
Bio-protocol, 6(16) (2016)
Cell Biology (2007)
Ruzica Bago et al.
The Biochemical journal, 463(3), 413-427 (2014-09-02)
The Vps34 (vacuolar protein sorting 34) class III PI3K (phosphoinositide 3-kinase) phosphorylates PtdIns (phosphatidylinositol) at endosomal membranes to generate PtdIns(3)P that regulates membrane trafficking processes via its ability to recruit a subset of proteins possessing PtdIns(3)P-binding PX (phox homology) and
Eric S Melby et al.
Langmuir : the ACS journal of surfaces and colloids, 34(36), 10793-10805 (2018-08-14)
Molecular understanding of the impact of nanomaterials on cell membranes is critical for the prediction of effects that span environmental exposures to nanoenabled therapies. Experimental and computational studies employing phospholipid bilayers as model systems for membranes have yielded important insights

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service