Skip to Content
Merck
All Photos(1)

Key Documents

810222C

Avanti

C6-NBD Glucosyl Ceramide

Avanti Research - A Croda Brand 810222C

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-D-glucosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H59N5O11
CAS Number:
Molecular Weight:
737.88
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810222C-250ug)
pkg of 1 × 1 mg (81022C-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand 810222C

concentration

0.25 mg/mL (810222C-250ug)

shipped in

dry ice

storage temp.

−20°C

General description

C6-NBD Glucosyl Ceramide is a fluorescent derivative of biologically active compound, glucosyl ceramide. Glucosyl ceramide belongs to the class of sphingolipids that are synthesized in the cis/medial-Golgi apparatus by the glycosylation of ceramide.

Application

C6-NBD Glucosyl Ceramide may be used as a substrate for glucocerebrosidase (GCase) activity assay in Drosophila melanogaster and mice.

Biochem/physiol Actions

Glucosylceramide is the precursor of different glycosphingolipids including gangliosides and sulfatides. Glucosyl ceramide is also known to be responsible for axon growth of neural cells. The accumulation of glucosyl ceramide is associated with an inherited lysosomal storage disorder, Gaucher′s disease.

Packaging

5 mL Amber Glass Screw Cap Vial (81022C-1MG)
5 mL Clear Glass Sealed Ampule (810222C-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhao et al.
Cellular and molecular life sciences : CMLS, 59(4), 694-707 (2002-05-23)
Gaucher disease is an autosomal recessive trait and the most common lysosomal storage disease. The pathogenesis evolves from the diminished activity of the lysosomal hydrolase, acid beta-glucosidase and the resultant accumulation of glucosylceramide within lysosomes. The pathogenic mechanisms are poorly
Yael Pewzner-Jung et al.
The Journal of biological chemistry, 285(14), 10902-10910 (2010-01-30)
Ceramide is an important lipid signaling molecule that plays critical roles in regulating cell behavior. Ceramide synthesis is surprisingly complex and is orchestrated by six mammalian ceramide synthases, each of which produces ceramides with restricted acyl chain lengths. We have
A Schwarz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(9), 2929-2938 (1997-05-01)
Sphingolipids (SLs) are important structural and regulatory components of neuronal plasma membranes. Previous studies using fumonisin B1, an inhibitor of the synthesis of ceramide, the precursor of all SLs, demonstrated that ceramide synthesis is required to sustain axonal growth in
Leonardo Astudillo et al.
Biochimie, 125, 267-280 (2015-11-20)
Sphingolipids represent a major class of lipids that are essential constituents of eukaryotic cells. They are predominantly located in plasma membrane microdomains, and play an important structural role in regulating membrane fluidity. They are also bioactive effectors involved in diverse
Or Cabasso et al.
Journal of clinical medicine, 8(9) (2019-09-12)
Gaucher disease (GD) results from mutations in the GBA1 gene, which encodes lysosomal glucocerebrosidase (GCase). The large number of mutations known to date in the gene lead to a heterogeneous disorder, which is divided into a non-neuronopathic, type 1 GD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service