Skip to Content
Merck
All Photos(2)

Key Documents

810164P

Avanti

16:0-12:0 NBD PG

Avanti Research - A Croda Brand 810164P, powder

Synonym(s):

1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H72N5O13P
CAS Number:
Molecular Weight:
862.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810164P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810164P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylglycerol (PG) is an anionic phospholipid(1 It is a predominant component of the cell membrane. Nitrobenzoxadiazole phosphoglycerol (NBD PG) is a fluorophore-labeled phospholipid.

Application

16:0-12:0 NBD PG or 1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt) may be used in microfluidic chip-based technology to demonstrate separation of fluorophore-labeled phospholipids.

Biochem/physiol Actions

Phosphatidylglycerol contributes to the biosynthesis of cardiolipin. It plays a vital role in photosynthetic activity by maintaining the structural photosynthetic apparatus.

Packaging

5 mL Amber Glass Screw Cap Vial (810164P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Plant & Cell Physiology null
Separation of phospholipids in microfluidic chip device: application to high-throughput screening assays for lipid-modifying enzymes
Lin S, et al.
Analytical biochemistry null
Separation of phospholipids in microfluidic chip device: application to high-throughput screening assays for lipid-modifying enzymes
Lin S, et al.
Analytical Biochemistry null
Furong Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 36(11), 2946-2953 (2020-02-26)
Phospholipase A2 is an important enzyme species which can widely be found in animals, plants, bacteria, and so on. A large number of studies have shown that phospholipase A2 is highly catalytic toward the lipids. Here, sum frequency generation (SFG)
Direct Evidence for Requirement of Phosphatidylglycerol in Photosystem II of Photosynthesis
Hagio M, et al.
Plant Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service