Skip to Content
Merck
All Photos(2)

Key Documents

810143C

Avanti

14:0 NBD PE

Avanti Research - A Croda Brand 810143C

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H70N5O11P
CAS Number:
Molecular Weight:
815.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810143C-1mg)
pkg of 5 × 1 mL (810143C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810143C

concentration

1 mg/mL (810143C-1mg)
1 mg/mL (810143C-5mg)

shipped in

dry ice

storage temp.

−20°C

Application

14:0 NBD PE is suitable for the preparation of phospholipid vesicles for lipid mixing assay. It is also suitable to use as fluorescent phospholipid derivative to study the effect of its exchange between β-lactoglobulin (β-LG) and phosphatidylcholine (POPC) bilayers.

Biochem/physiol Actions

NBD PE (1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl)) can be used liposome preparation. In which, it might serve as a measure of steady-state fluorescence anisotropy. This might be useful to analyse the mitochondrial membrane structure.

Packaging

5 mL Amber Glass Screw Cap Vial (810143C-1mg)
5 mL Amber Glass Screw Cap Vial (810143C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro Membrane Interaction and Liposome Fusion Assays Using Recombinant Hepatitis C Virus Envelope Protein E2
Yelamos B, et al.
Bio-protocol, 8(23), 82-89 (2018)
Cardiolipin and phosphatidylethanolamine role in dibucaine interaction with the mitochondrial membrane.
Lopes SC, et al.
Biochimica et Biophysica Acta - Biomembranes, 186(6), 1152-1161 (2019)
Binding of phospholipids to ?-Lactoglobulin and their transfer to lipid bilayers
P.A.T.Martins, et al.
Biochimica et Biophysica Acta - Biomembranes, 1778(5), 1308-1315 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service