Skip to Content
Merck
All Photos(1)

Key Documents

241601A

Avanti

L-α-Phosphatidylcholine

60% (Egg), Avanti Polar Lipids, Paste

Synonym(s):

L-α-Phosphatidylcholine (Egg, Chicken-60%) (Total Egg Phosphatide Extract)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Product Name

Egg PC (60%), Avanti Research - A Croda Brand

form

paste

packaging

pkg of 1 × 1 g (241601A-1g)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is a critical component of the mucosal layer of the colon and is the most common phospholipid in mammalian membranes. It is a strong bilayer-forming lipid.
Total Egg Extract is prepared by extracting powdered egg yolk with methanol. The methanol extract is partitioned with chloroform and deionized water, and the chloroform phase containing the organic soluble lipids is concentrated.

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance. It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.

Packaging

60 mL Amber Wide Mouth Screw Cap Glass Bottle (241601A-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book (2017)
The Membranes of Cells (2016)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service