Z742795
Kern EMB series precision balances
Kern EMB 200-3, weighing capacity 200 g, accuracy: 0.001 g, precision: ±0.005 g
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
All Photos(1)
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Kern EMB 200-3
weighing capacity measuring range
200 g, accuracy: 0.001 g, precision: ±0.005 g
Related Categories
General description
Entry level laboratory balance with tremendous weighing performance
- Simple and convenient 2-key operation, making them ideal for use in schools and universities
- Tare function facilitates formulation work
- Particularly flat design
- Secure and non-slip positioning with rubber feet
- Ready for use: Batteries included. AUTO-OFF function to preserve the batteries, can be switched off
Choose from one of the most recent versions:
Certificates of Analysis (COA)