Skip to Content
Merck
All Photos(2)

Key Documents

Z676519

ART® barrier tip

maximum volume 10 μL, Tip design ART 10, Hinged rack, low retention: no

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
10 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
endotoxin free
low retention: no
Hinged rack
Tip design ART 10
packaging:
pack of 96 ea (Hinged rack)
manufacturer/tradename:
Molecular BioProducts 2139-HR
Pricing and availability is not currently available.

material

colorless
colorless tip

sterility

sterile

feature

DNase free
RNase free
barrier
endotoxin free
low retention: no
Hinged rack
Tip design ART 10

packaging

pack of 96 ea (Hinged rack)

manufacturer/tradename

Molecular BioProducts 2139-HR

technique(s)

PCR: suitable

maximum volume

10 μL

Looking for similar products? Visit Product Comparison Guide

General description

The self-sealing ART barrier tips are supplied in environmentally friendly packaging. The sterile, barrier tip reload system combines individually packaged blister tubs made of recycled plastic and new hinged racks. The push down blister tub easily inserts the ART barrier tips into the hinged racks providing stackable, space saving racks that offer easy, one-handed use. Empty hinged rack part numbers allow for easy conversion to the ART Reload System.

Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required.

Available as reload inserts consisting of sterile pre-loaded blister tubs of 96 tips with or without hinged racks. The empty hinged racks are also available separately.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service