Key Documents
Z616893
Vacuum accessories for Buchi® vacuum pumps
Woulff flask
About This Item
UNSPSC Code:
41105101
NACRES:
NB.43
packaging
pack of 1 ea
manufacturer/tradename
Büchi 047170
General description
This optional Woulff flask may be connected in front of pump as a vacuum reservoir to protect pump from liquids accidentally sucked into vacuum line. Recommended in stand-alone systems using a V-700/710 pump. Includes retaining ring for V-700.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)