Key Documents
Z417394
Sigma-Aldrich
Aldrich® System 45 three-hole delivery cap
PTFE, thread for 45 mm, pkg of 1 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
PTFE
packaging
pkg of 1 ea
fitting
thread for 45 mm
General description
Cap has 3 holes with 1/4-28 threads at the top for use with 1/8 in. O.D. tubing. No need to precut tubing to reach the reservoir bottom, just a quick quarter turn of the lock nut (from outside the reservoir) allows positioning of PTFE tubing at any depth. Thread size: 45 mm.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
Choose from one of the most recent versions:
Certificates of Analysis (COA)