Skip to Content
Merck
All Photos(2)

Documents

Z416436

Supelco

EZSafe® flash-chromatography assembly

capacity 2,000 mL, With fritted disc (porosity C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NC.05

grade

for chromatography

technique(s)

LPLC: suitable

capacity

2,000 mL

Looking for similar products? Visit Product Comparison Guide

General description

These columns are improved low-pressure columns (max. pressure 7psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy.
  • Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE
  • GL-45 screw-thread connections ensure positive reliable connections
  • Eliminates frozen PTFE and glass plugs
  • Eliminates grease
  • Choose between standard columns or columns with a fritted disc

The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.

Legal Information

EZ Safe is a registered trademark of Gerresheimer Glass, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service